Skip to Content
MilliporeSigma

Skip To

90769

(+)-Abscisic acid

analytical standard

Synonym(s):

(S)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-(2Z,4E)-pentadienoic acid, ABA, Dormin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
UNSPSC Code:
12352106
NACRES:
NA.24
PubChem Substance ID:
EC Number:
244-319-5
Beilstein/REAXYS Number:
2698956
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.0% (HPLC)

optical activity

[α]/D 415±10°, c = 0.1 in ethanol

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

CC(\C=C\[C@@]1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O

InChI

1S/C15H20O4/c1-10(7-13(17)18)5-6-15(19)11(2)8-12(16)9-14(15,3)4/h5-8,19H,9H2,1-4H3,(H,17,18)/b6-5+,10-7-/t15-/m1/s1

InChI key

JLIDBLDQVAYHNE-YKALOCIXSA-N

Biochem/physiol Actions

Plant hormone and growth regulator that is involved in several physiological mechanisms including seed dormancy, leaf abscission, stomatal movement, and plant stress responses. Through complex interactions with several intracellular signaling systems, it can regulate the expression of hundreds of plant genes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A4906862169PHL84142
grade

analytical standard

grade

-

grade

-

grade

primary reference standard

format

neat

format

-

format

-

format

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-

assay

≥98.0% (HPLC)

assay

≥98% (HPLC)

assay

98%

assay

≥98.0% (HPLC)

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

-

optical activity

[α]/D 415±10°, c = 0.1 in ethanol

optical activity

-

optical activity

-

optical activity

-


Still not finding the right product?

Explore all of our products under (+)-Abscisic acid


signalword

Warning

hcodes

pictograms

Environment

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. where should abscisic acid be solubilised for use in a plate germination test?

    1 answer
    1. The solubility of this product is tested in ethanol at 10 mg/mL. For plant tissue culture applications, 1N NaOH or 1N KOH is typically used.

      Helpful?

Reviews

No rating value

Active Filters