Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

516566

Sigma-Aldrich

PPARγ Antagonist III, G3335

The PPARγ Antagonist III, G3335, also referenced under CAS 36099-95-3, controls the biological activity of PPARγ.

Synonym(s):

PPARγ Antagonist III, G3335, H-Trp-Glu-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O5
CAS Number:
Molecular Weight:
333.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 30 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[nH]1c2c(c(c1)C[C@H](N)C(=O)N[C@@H](CCC(=O)O)C(=O)O)cccc2

InChI

1S/C16H19N3O5/c17-11(7-9-8-18-12-4-2-1-3-10(9)12)15(22)19-13(16(23)24)5-6-14(20)21/h1-4,8,11,13,18H,5-7,17H2,(H,19,22)(H,20,21)(H,23,24)/t11-,13-/m0/s1

InChI key

PWIQCLSQVQBOQV-AAEUAGOBSA-N

General description

A cell-permeable dipeptide that acts as a selective and reversible PPARγ antagonist (KD = ~ 8 µM). Shown to inhibit the agonist activity of Rosiglitazone (100 nM) in a dose-dependent manner (IC50 = 31.9 µM) in transfected COS-7 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PPARγ
Product does not compete with ATP.
Reversible: yes
kd = ~ 8 µM as PPARγ antagonist

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Preparation Note

Addition of a small amount of acid or base may be required for complete solubilization.

Reconstitution

Following reconstitution aliquot freeze at -20°C. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ye, F., et al. 2006. Chembiochem7, 74.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service