Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

512732

Sigma-Aldrich

Parthenolide, Tanacetum parthenium

A cell-permeable sesquiterpene lactone with anti-inflammatory, antisecretory, and spasmolytic properties.

Synonym(s):

Parthenolide, Tanacetum parthenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O3
CAS Number:
Molecular Weight:
248.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL
ethanol: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O1[C@H]2[C@H]3OC(=O)C(=C)[C@@H]3CC\C(=C/CC[C@]21C)\C

InChI

1S/C15H20O3/c1-9-5-4-8-15(3)13(18-15)12-11(7-6-9)10(2)14(16)17-12/h5,11-13H,2,4,6-8H2,1,3H3/b9-5-/t11-,12-,13-,15+/m0/s1

InChI key

KTEXNACQROZXEV-QLIGOWBFSA-N

General description

A cell-permeable sesquiterpene lactone with anti-inflammatory, antisecretory, and spasmolytic properties. Inhibits the expression of COX-2 and proinflammatory cytokines (TNF-α and IL-1) in LPS-stimulated macrophages. Also inhibits NF-κB and activation of MAP kinase.
A sesquiterpene lactone with anti-inflammatory, antisecretory, and spasmolytic properties. Inhibits the expression of COX-2 and proinflammatory cytokines (TNF-α and IL-1) in LPS-stimulated macrophages. Also inhibits NF-κB and activation of MAP kinase.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
the expression of COX-2 and proinflammatory cytokines (TNF-α and IL-1) in LPS-stimulated macrophages
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Other Notes

Wong, H.R., et al. 1999. Biochem. Biophys. Res. Comm.262, 375.
Bork, P.M., et al. 1997. FEBS Lett. 402, 85.
Hwang, D., et al. 1996. Biochem. Biophys. Res. Commun. 226, 810.
Barsby, R.W., et al. 1992. J. Pharm. Pharmacol. 44, 737.
Sumner, H., et al. 1992. Biochem. Pharmacol. 43, 2313.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yangming Xiao et al.
Journal of immunology (Baltimore, Md. : 1950), 174(3), 1701-1708 (2005-01-22)
Chlamydia trachomatis, an obligate intracellular bacterial species, is known to inhibit host cell apoptosis. However, the chlamydial antiapoptotic mechanism is still not clear. Because NF-kappaB activation is antiapoptotic, we tested the potential role of NF-kappaB activation in chlamydial antiapoptotic activity
Seok-Man Kim et al.
Frontiers in cell and developmental biology, 12, 1398176-1398176 (2024-07-26)
Phthalate esters (PAEs) are primary plasticizers and endocrine-disrupting chemicals (EDCs) that are extensively used in numerous everyday consumer products. Although the adverse effects of single PAEs have been studied, our understanding of the effect of multiple phthalate exposure on male
M Marquina et al.
The British journal of dermatology, 159(1), 68-76 (2008-05-15)
Pemphigus vulgaris (PV) is a blistering autoimmune disease characterized by IgG autoantibodies against desmoglein 3. Nitric oxide synthases (NOS) may contribute to the increase of inflammation in tissues by the generation of nitrotyrosine residues (NTR). To investigate whether the production

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service