Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.08227

Sigma-Aldrich

Paclitaxel, Taxus sp.

InSolution, ≥97%, 25 mM in DMSO

Synonym(s):

InSolution Paclitaxel, Taxus sp., TAXOL® InSolution, Baccatin III N-benzyl-β-phenylisoserine Ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H51NO14
CAS Number:
Molecular Weight:
853.91
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

N([C@H]([C@@H](O)C(=O)O[C@H]3C[C@]4([C@H](C6[C@]7([C@H](OC7)C[C@@H]([C@]6(C(=O)[C@@H](C(=C3C)C4(C)C)OC(=O)C)C)O)OC(=O)C)OC(=O)c5ccccc5)O)c2ccccc2)C(=O)c1ccccc1

InChI

1S/C47H51NO14/c1-25-31(60-43(56)36(52)35(28-16-10-7-11-17-28)48-41(54)29-18-12-8-13-19-29)23-47(57)40(61-42(55)30-20-14-9-15-21-30)38-45(6,32(51)22-33-46(38,24-58-33)62-27(3)50)39(53)37(59-26(2)49)34(25)44(47,4)5/h7-21,31-33,35-38,40,51-52,57H,22-24H2,1-6H3,(H,48,54)/t31-,32-,33+,35-,36+,37+,38?,40-,45+,46-,47+/m0/s1

InChI key

RCINICONZNJXQF-VAZQATRQSA-N

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 25 mM (10 mg/468 µL) sterile-filtered solution of Paclitaxel, Taxus sp. (Cat. No. 580555) in DMSO.

Other Notes

Derry, W.B., et al. 1995. Biochemistry34, 2203.
Milas, L., et al. 1995. Cancer Chemother. Pharmacol.35, 297.
Donaldson, K.L., et al. 1994. Int. J. Cancer57, 847.
Hornback, N.B., et al. 1994. In Vivo8, 819.
Manthey, C.L., et al. 1992. J. Immunol.149, 2459.
Rowinsky, E.K., et al. 1992. Semin. Oncol. 19, 646.
Ding, A.H., et al. 1990. Science248, 370.
Mogensen, M.M. and Tucker, J.B. 1990. J. Cell Sci.97, 101.
Slichenmeyer, W.J. and Von Hoff, D.D. 1990. J. Clin. Pharmacol.30, 770.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Taxol is a registered trademark of Bristol-Myers Squibb Co.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service