Skip to Content
MilliporeSigma
All Photos(1)

Documents

5.04776

Sigma-Aldrich

Cytochalasin D, Zygosporium mansonii

InSolution, ≥98%, 10 mM in DMSO, A cell-permeable fungal toxin

Synonym(s):

InSolution Cytochalasin D, Zygosporium mansonii, Lygosporin A, Zygosporin A, NSC 209835

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H37NO6
CAS Number:
Molecular Weight:
507.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

InChI

1S/C30H37NO6/c1-17-10-9-13-22-26(33)19(3)18(2)25-23(16-21-11-7-6-8-12-21)31-28(35)30(22,25)24(37-20(4)32)14-15-29(5,36)27(17)34/h6-9,11-15,17-18,22-26,33,36H,3,10,16H2,1-2,4-5H3,(H,31,35)/b13-9+,15-14+/t17-,18+,22-,23-,24+,25-,26+,29+,30+/m0/s1

InChI key

SDZRWUKZFQQKKV-JHADDHBZSA-N

General description

A cell-permeable fungal toxin that is approximately 10 times more potent than cytochalasin B (>Cat. No. 250233) in inhibiting actin filament function. Does not inhibit monosaccharide transport across cell membranes. Inhibits cytoplasmic division by blocking the formation of contractile microfilaments at 10 µM. Inactivates low conductance K+ channels. Also modulates CD4 cross-linking in T lymphocytes and increases intracellular Ca2+ levels. Cytochalasin D also exhibits antibiotic and anti-tumor properties.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
microfilaments
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (1 mg/197 µL) sterile-filtered solution of Cytochalasin D, Zygosporium mansonii (Cat. No. 250255) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C.

Other Notes

Sasaki, H., et al. 1995. Proc. Natl. Acad. Sci. USA92, 2026.
Blankson, H., et al. 1995. Exp. Cell Res.218, 522.
Melamed, I., et al. 1995. Biochem. Biophys. Res. Commun.209, 1102.
Verschueren, H., et al. 1995. Eur. J. Cell Biol.66, 335.
Aszalos, A., et al. 1994. Cell Immunol. 157, 81.

Wang, W.H., et al. 1994. Am. J. Physiol. 267, F592.

Betina, V., and Micekova, D. 1972. Chem. Abstr.77, 160508q.
Katagiri, K., and Matsuura, S. 1971. J. Antibiot. 24, 722.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sylwia Jones et al.
Scientific reports, 10(1), 663-663 (2020-01-22)
Antibody combinations targeting cell surface receptors are a new modality of cancer therapy. The trafficking and signalling mechanisms regulated by such therapeutics are not fully understood but could underlie differential tumour responses. We explored EGFR trafficking upon treatment with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service