Skip to Content
MilliporeSigma
All Photos(1)

Documents

475823-M

Sigma-Aldrich

β₂-Microglobulin, Human Urine

Synonym(s):

β₂M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.28

Assay

≥98% (SDS-PAGE)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

Native β2-microglobulin from human urine. A useful marker for the determination of AIDS progression in HIV-infected subjects. Increased levels may also indicate increased proliferation of tumor masses.
Native β2-microglobulin from human urine. Single chain polypeptide that is the constant component of Class I MHC, which is located on most nucleated cells. It is also a normal constituent of serum and urine. Increased levels of this protein in the serum are used as a marker for impairments of renal function and a variety of lymphoproliferative diseases.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 150 mM NaCl, 20 mM phosphate buffer, pH 7.3.

Preparation Note

Prepared from urine that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, product can be stored at 4°C for up to 6 months.

Other Notes

Zabay, J.M., et al. 1995. J. Acquired Immune Defic. Syndr. Hum. Retrovirol. 8, 266.
Mady, B.J., et al. 1991. J. Immunol.147, 3139.
Lampson, L.A. 1990. J. Immunol. 144, 512.
Calzia, R., et al. 1989. Adv. Exp. Med. Biol.257, 225.
Ninomiya, Y., and Arakawa, M. 1989. Diabetes Res.10, 129.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service