Skip to Content
MilliporeSigma

4500-M

L-Methionine

An essential amino acid, useful as a methyl group donor.

Synonym(s):

L-Methionine, Met

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H11NO2S
CAS Number:
Molecular Weight:
149.21
UNSPSC Code:
12352209
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

RTECS - PD0457000
Merck USA index - 14, 5975

assay

≥98.5% (by assay)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

colorless to white

solubility

water: 1 mg/mL

cation traces

heavy metals: ≤20 ppm

storage temp.

10-30°C

SMILES string

S(CCC(N)C(=O)O)C

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

InChI key

FFEARJCKVFRZRR-UHFFFAOYSA-N

General description

An essential amino acid, useful as a methyl group donor.
An essential amino acid. Useful as a methyl group donor.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under L-Methionine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library