Skip to Content
MilliporeSigma
L-Methionine SAJ special grade, ≥98.5%

L-Methionine

L-Methionine SAJ special grade, ≥98.5%
Synonym(s):
(S)-2-Amino-4-(methylmercapto)butyric acid, L-2-Amino-4-(methylthio)butanoic acid, L-Methionine, L-Methionine
Linear Formula:
CH3SCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
149.21
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Amino Acid
API Family
Reference Material Type
Special Grade
Technique