Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444289

Sigma-Aldrich

Marimastat

An orally active peptidyl hydroxamate-based broad-spectrum MMP inhibitor that targets both the substrate binding site and the active-site Zn2+.

Synonym(s):

Marimastat, (2S,3R)-N⁴-((1S)-2,2-Dimethyl-1-((methylamino)carbonyl)propyl)-N¹,2-hydroxy-3-(2-methylpropyl)butanediamide, BB-2516

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H29N3O5
CAS Number:
Molecular Weight:
331.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (NMR)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 15 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C15H29N3O5/c1-8(2)7-9(10(19)13(21)18-23)12(20)17-11(14(22)16-6)15(3,4)5/h8-11,19,23H,7H2,1-6H3,(H,16,22)(H,17,20)(H,18,21)/t9-,10+,11-/m1/s1

InChI key

OCSMOTCMPXTDND-OUAUKWLOSA-N

General description

An orally active peptidyl hydroxamate-based broad-spectrum MMP inhibitor (IC50 in nM = 5, 6, 200, 20, 2, 1.8 and 3.8 for MMP-1, -2, -3, -7, -8, -14 and TACE, respectively) that targets both the substrate binding site and the active-site Zn2+. Marimastate is widely used in studying the involvement of MMPs in various cellular and pathological processes both in vitro and in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Hansen, H.P., et al. 2002. Int. J. Cancer98, 210.
Tsuji, F., et al. 2002. Cytokine17, 294.
Barlaam, B., et al. 1999. J. Med. Chem.42, 4890.
Whittaker, M., et al. 1999. Chem. Rev.99, 2735.
Rasmussen, H.S., and McCann, P.P. 1997. Pharmacol. Ther.75, 69.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service