Skip to Content
MilliporeSigma
All Photos(1)

Documents

437647

Sigma-Aldrich

Lipoproteins, Very Low Density, Human Plasma

Synonym(s):

VLDL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

Assay

≥95% (of total lipoprotein content, electrophoresis)

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

shipped in

wet ice

storage temp.

2-8°C

General description

A 5 mg vial contains ~1 mg of protein.
Native very low density lipoproteins from human plasma. VLDL transports hepatic synthesized triglycerides and cholesterol. May counteract the inhibitory effect of glucocorticoids on arachidonic acid release and prostaglandin I2 formation in vascular smooth muscle cells. (Composition: 88-95% lipid; 5-12% protein. Note: 5 mg vial contains 1 mg of protein.)
Native very low density lipoproteins from human plasma. VLDL transports liver-synthesized triglycerides and cholesterol. May counteract the inhibitory effect of glucocorticoids on arachidonic acid release and prostaglandin I2 formation in vascular smooth muscle cells. Composition: 88-95% lipid; 5-12% protein.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Other Notes

Petrichenko, I., et al. 1993. Biochim. Biophys. Acta1166, 183.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wojciech Paslawski et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(30), 15226-15235 (2019-07-05)
The progressive accumulation, aggregation, and spread of α-synuclein (αSN) are common hallmarks of Parkinson's disease (PD) pathology. Moreover, numerous proteins interact with αSN species, influencing its toxicity in the brain. In the present study, we extended analyses of αSN-interacting proteins
Wei Shao et al.
The Journal of biological chemistry, 295(52), 18284-18300 (2020-10-29)
Oxygen regulates hypoxia-inducible factor (HIF) transcription factors to control cell metabolism, erythrogenesis, and angiogenesis. Whereas much has been elucidated about how oxygen regulates HIF, whether lipids affect HIF activity is un-known. Here, using cultured cells and two animal models, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service