Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

401105

Sigma-Aldrich

Importazole

A cell-permeable diaminoquinazoline compound that selectively blocks importin-β-mediated nuclear import of NLS bearing cargos in a reversible manner.

Synonym(s):

Importazole, Importin-β Inhibitor, N-(1-Phenylethyl)-2-pyrrolidin-1-ylquinazolin-4-amine, IPZ, N-(1-Phenylethyl)-2-pyrrolidin-1-ylquinazolin-4-amine, IPZ, Importin-β Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N4
CAS Number:
Molecular Weight:
318.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(C1=CC=CC=C1)NC2=NC(=NC3=CC=CC=C32)N4CCCC4

InChI key

HKGJEZIGDHFJFL-UHFFFAOYSA-N

General description

A cell-permeable diaminoquinazoline compound that selectively blocks importin-β-mediated nuclear import of NLS- (nuclear localization signal) bearing cargos (IC50 = 15 µM against Ca+2-induced NFAT nuclear import in HEK293 cells) in a reversible manner without affecting transportin-dependent nuclear import of M9 signal-containing cargos or CRM1-mediated nuclear export. IPZ affects, but not blocks, importin-β-RanGTP interaction, likely by inducing a conformational change upon direct importin-β binding. IPZ treatment impairs cellular mitosis and is cytotoxic to HeLa cells (50% and >90% killing in 24 h, respectively, with 22.5 and 60 µM IPZ).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Soderholm, J.F., et al. 2011. ACS Chem. Biol.6, 700.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Luo et al.
Molecular therapy. Methods & clinical development, 20, 551-558 (2021-03-06)
The anti-inflammatory molecule annexin A1 (ANXA1) determines the ultimate fate of retinal ganglion cell (RGC) in glaucoma. Cytoplasmic and extracellular ANXA1 facilitate resolution of inflammation. However, the nuclear translocation of ANXA1 induces RGC apoptosis in a murine glaucoma model, and
Yu-Ching Dai et al.
Journal of virology, 94(3) (2019-11-07)
Epstein-Barr virus (EBV) genomic DNA is replicated and packaged into procapsids in the nucleus to form nucleocapsids, which are then transported into the cytoplasm for tegumentation and final maturation. The process is facilitated by the coordination of the viral nuclear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service