Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39468-M

Millipore

Tricine

ULTROL® Grade, ≥99% (titration), zwitterionic buffer useful in pH range 7.4-8.8

Synonym(s):

Tricine, ULTROL® Grade, N-[ tris(Hydroxymethyl)methyl]glycine, N-[tris(Hydroxymethyl)methyl]glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO5
CAS Number:
Molecular Weight:
179.17
MDL number:
UNSPSC Code:
12161705
NACRES:
NA.25

description

Merck USA index - 14, 9651

Quality Level

Assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

pKa (20 °C)

8.2

solubility

water: 20%

cation traces

heavy metals: ≤1 ppm

storage temp.

10-30°C

SMILES string

[N+H2](C(CO)(CO)CO)CC(=O)[O-]

InChI

1S/C6H13NO5/c8-2-6(3-9,4-10)7-1-5(11)12/h7-10H,1-4H2,(H,11,12)

InChI key

SEQKRHFRPICQDD-UHFFFAOYSA-N

General description

A zwitterionic amino acid commonly useful in the pH range of 7.4-8.8. Has a pKa 8.05 at 25°C.
A zwitterionic buffer useful in the pH range of 7.4-8.8. Has a pKa 8.05 at 25°C.

Application


  • Rational Design and Comparison of Novel (99m)Tc-Labeled FAPI Dimers for Visualization of Multiple Tumor Types.: This study focuses on the design and evaluation of novel radiolabeled compounds for imaging various tumor types. Tricine is used as a buffer component to stabilize the radioactive isotopes, enhancing the imaging quality for precise tumor localization (Meng et al., 2024).

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service