Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

385931

Sigma-Aldrich

Hyaluronidase, Bovine Testes

Synonym(s):

Hyaluronidase, Bovine Testes

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

conjugate

conjugate (Glucosaminoglycan)

Quality Level

description

Merck USA index - 14, 4758

form

lyophilized

specific activity

≥2500 units/mg dry wt.

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

−20°C

General description

Native hyaluronidase from bovine testes. Catalyzes the hydrolysis of 1,4 linkages between N-acetylhexosamine and D-glucuronate residues in chondroitin, chondroitin-4-sulfate, chondroitin-6-sulfate, and hyaluronic acid.
Native hyaluronidase from bovine testes. Hyaluronic acid degrading endoglycosidase that catalyzes the hydrolysis of 1,4 linkages between N-acetylhexosamine and D-glucuronate residues in chondroitin, chondroitin-4-sulfate, chondroitin-6-sulfate, and hyaluronic acid.
Note: 1 KU = 1000 units.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will cause the same turbidity reduction as 1.0 unit of International Standard preparation.

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C.
Reconstitute in 20 mM sodium phosphate, pH 6.9, 0.45% NaCl, and 0.01% BSA.

Other Notes

Lokeshwar, V.B., et al. 2002. J. Biol. Chem.277, 33654.
Humphrey, I.H. 1957. Bull. WHO16, 291.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H HUMPHREY
Bulletin of the World Health Organization, 16(2), 291-294 (1957-01-01)
The International Standard for Hyaluronidase is derived from a blend of four batches of bovine testicular hyaluronidase which had been purified so as to contain approximately 400 units per milligram. The material was mixed with beta-lactose, and lyophilized. Tablets were
Vinata B Lokeshwar et al.
The Journal of biological chemistry, 277(37), 33654-33663 (2002-06-27)
Hyaluronidase is a hyaluronic acid-degrading endoglycosidase that is present in many toxins and the levels of which are elevated in cancer. Increased concentration of HYAL1-type hyaluronidase correlates with tumor progression and is a marker for grade (G) 2 or 3

Articles

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

Explore the role of hyaluronan, hyaluronic acid structure, hyaluronan synthesis & degradation, and more. Find GAGs, hydrogels, and scaffold kits.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service