Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

361556

Sigma-Aldrich

GSK-3 Inhibitor IX, Control, MeBIO

The GSK-3 Inhibitor IX, Control, MeBIO, also referenced under CAS 710323-61-8, controls the biological activity of GSK-3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

GSK-3 Inhibitor IX, Control, MeBIO, 1-Methyl-BIO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12BrN3O2
CAS Number:
Molecular Weight:
370.20
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable N-methylated analog of GSK-3 Inhibitor IX, BIO (Cat. No. 361550) that serves as a relevant kinase inactive control (IC50 >92 µM for Cdk1/B, >100 µM for Cdk5/p25, and GSK-3α/β). Acts as a potent ligand for aryl hydrocarbon receptor (AhR; EC50 = 20 nM and 93 nM for yeast and hepatoma reporter systems, respectively) and displays antiproliferative properties.
A cell-permeable N-methylated analog of GSK-3 Inhibitor IX, BIO (Cat. No. 361550) that serves as an inactive control for Cdk1/cyclin B, Cdk5/p25, and GSK-3α/β (IC50 >92 µM for Cdk1/cyclin B, >100 µM for Cdk5/p25, and GSK-3α/β). Acts as a potent ligand for aryl hydrocarbon receptor (AhR; EC50 = 20 nM and 93 nM for yeast and hepatoma reporter systems, respectively) and displays antiproliferative properties.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Relevant kinase inactive control
Product does not compete with ATP.
Reversible: no
Target IC50: >92 µM for Cdk1/B, >100 µM for Cdk5/p25, and GSK-3α/β

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verónica Uribe-Etxebarria et al.
Cells, 9(3) (2020-03-12)
Dental pulp stem cells (DPSCs) from adult teeth show the expression of a very complete repertoire of stem pluripotency core factors and a high plasticity for cell reprogramming. Canonical Wnt and Notch signaling pathways regulate stemness and the expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service