345836
Genistin
An isoflavone glycoside found in soy-based food products.
Synonym(s):
Genistin, Genistein, 7-O-β-D-Glucopyranoside
About This Item
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white to light yellow
solubility
DMSO: 10 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)Oc2cc3[o]cc([c](c3c(c2)O)=O)c4ccc(cc4)O
InChI
1S/C21H20O10/c22-7-15-18(26)19(27)20(28)21(31-15)30-11-5-13(24)16-14(6-11)29-8-12(17(16)25)9-1-3-10(23)4-2-9/h1-6,8,15,18-24,26-28H,7H2/t15-,18-,19+,20-,21-/m1/s1
InChI key
ZCOLJUOHXJRHDI-CMWLGVBASA-N
General description
Biochem/physiol Actions
A negative control for Genistein
Packaging
Warning
Reconstitution
Other Notes
Kajiya, H., et al. 2000. J. Cell. Physiol.183, 83.
Hamakawa, T., et al. 1999. J. Neurosci. 19, 9306.
Wang, H., and Murphy, P.A. 1994. Agric. Food Chem. 42, 1666.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service