Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345834

Sigma-Aldrich

Genistein, Soybean

A cell-permeable, reversible, substrate competitive inhibitor of protein tyrosine kinases, including autophosphorylation of epidermal growth factor receptor kinase (IC₅₀ = 2.6 µM).

Synonym(s):

Genistein, Soybean, 4ʹ,5,7-Trihydroxyisoflavone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 4391

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 200 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[o]1c2c([c](c(c1)c3ccc(cc3)O)=O)c(cc(c2)O)O

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-13-6-10(17)5-12(18)14(13)15(11)19/h1-7,16-18H

InChI key

TZBJGXHYKVUXJN-UHFFFAOYSA-N

General description

A cell-permeable, reversible, substrate competitive inhibitor of protein tyrosine kinases, including autophosphorylation of epidermal growth factor receptor kinase (IC50 = 2.6 µM). The inhibition is competitive with respect to ATP and non-competitive with respect to the phosphate acceptor. Inhibits tumor cell proliferation and induces tumor cell differentiation. Inhibits exogenous substrate phosphorylation by EGFR kinase and pp60v-src with similar potencies (IC50 = 20-25 µM). Has only a trivial effect on PKC and PKA (IC50 >350 µM). Produces cell cycle arrest and apoptosis in Jurkat T-leukemia cells. Prevents anti-CD3 monoclonal antibody-induced thymic apoptosis. Also inhibits topoisomerase II activity in vitro. Blocks oxidative DNA damage in vitro.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EGFR kinase
Product does not compete with ATP.
Reversible: yes
Target IC50: 2.6 µM against autophosphorylation of epidermal growth factor receptor kinase

Warning

Toxicity: Standard Handling (A)

Other Notes

Constantinou, A., and Huberman, E. 1995. Proc. Soc. Exp. Biol. Med. 208, 109.
Wei, H., et al. 1995. Proc. Soc. Exp. Biol. Med. 208, 124.
Wei, H., et al. 1995. Carcinogenesis17, 73.
Kobayashi, S., et al. 1994. J. Biol. Chem.269, 9011.
Migita, K., et al. 1994. J. Immunol.153, 3457.
Spinozzi, F., et al. 1994. Leuk. Res.18, 431.
Dhar, A., et al. 1990. Mol. Pharmacol.37, 519.
Hill, T.D., et al. 1990. Science248, 1660.
Dean, N.M., et al. 1989. Biochem. Biophys. Res. Commun.165, 795.
Akiyama, T., et al. 1987. J. Biol. Chem.262, 5592.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shanqing Yi et al.
Folia histochemica et cytobiologica, 59(1), 49-56 (2021-02-20)
Alzheimer's disease (AD), a very common neurodegenerative disorder, is mainly characterized by the deposition of b-amyloid protein (Ab) and extensive neuronal cell death. Currently, there are no satisfactory therapeutic approaches for AD. Although neuroprotective effects of genistein against Ab-induced toxicity
Wei Zhang et al.
Development (Cambridge, England), 149(21) (2022-10-06)
Migration of myoblasts derived from the occipital somites is essential for tongue morphogenesis. However, the molecular mechanisms of myoblast migration remain elusive. In this study, we report that deletion of Isl1 in the mouse mandibular epithelium leads to aglossia due
Chenyao Wang et al.
The EMBO journal, 39(22), e104106-e104106 (2020-09-15)
STING (STimulator of INterferon Genes) mediates protective cellular response to microbial infection and tissue damage, but its aberrant activation can lead to autoinflammatory diseases. Upon ligand stimulation, the endoplasmic reticulum (ER) protein STING translocates to endosomes for induction of interferon
Blanche C Ip et al.
Toxicological sciences : an official journal of the Society of Toxicology, 186(1), 29-42 (2021-12-23)
Humans are consistently exposed to thousands of untested chemicals that have been detected in the follicular fluid of the ovaries, and can disrupt reproductive health. Human granulosa cells (GCs) are the functional unit of the ovarian follicle with steroidogenic and
J C Norman et al.
FEBS letters, 484(3), 179-183 (2000-11-18)
Aggregation by immune complexes of receptors specific for the Fc region of IgG results in their internalisation and disposal by trafficking to lysosomes. We show here that internalisation of FcgammaRI by IFN-gamma treated U937 cells following receptor aggregation by cross-linking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service