Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345752

Sigma-Aldrich

Ganglioside GD₃, Disialo, Diammonium Salt, Bovine Buttermilk

Has been characterized as one of the most important tumor-specific antigens in individuals with malignant melanoma.

Synonym(s):

Ganglioside GD₃, Disialo, Diammonium Salt, Bovine Buttermilk, Neu5Acα2,8Neu5Acα2,3Galβ1,4Glcβ1,1 Ceramide, 2NH₄

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H123N3O30 · 2NH4
Molecular Weight:
1522.80
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 1 mg/mL
chloroform/methanol (2:1): soluble

shipped in

ambient

storage temp.

−20°C

General description

Has been characterized as one of the most important tumor-specific antigens in individuals with malignant melanoma. Reported to disrupt mitochondrial transmembrane potential and induce apoptosis in tumor cells by a caspase-independent mechanism. Antibodies to GD3 can induce partial remission of tumor growth in animals and in humans via enhancement of cytotoxic and proliferative responses of lymphocytes. M.W. 1577.0 (based on tricosanoyl - 23:0).

Warning

Toxicity: Irritant (B)

Other Notes

DeMaria, R., et al. 1997. Science 277, 1652.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumeyye Cavdarli et al.
Cells, 10(6) (2021-07-03)
The O-acetylated form of GD2, almost exclusively expressed in cancerous tissues, is considered to be a promising therapeutic target for neuroectoderm-derived tumors, especially for breast cancer. Our recent data have shown that 9-O-acetylated GD2 (9-OAcGD2) is the major O-acetylated ganglioside

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service