Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345744

Sigma-Aldrich

Ganglioside GT1b, Trisialo, Triammonium Salt, Bovine Brain

Inhibits Con A-stimulated mitogenesis in murine T cells.

Synonym(s):

Ganglioside GT1b, Trisialo, Triammonium Salt, Bovine Brain, Neu5Acα2,3Galβ1,3GalNAcβ1,4(Neu5Acα2,8Neu5Acα2,3)Galβ1,4Glcβ1,1 Ceramide, 3NH₄

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C95H162N5O47 · 3NH4
Molecular Weight:
2180.42
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

ethanol: soluble
methanol: soluble

shipped in

ambient

storage temp.

−20°C

General description

Inhibits Con A-stimulated mitogenesis in murine T cells. Lymphocyte marker in cancer patients. Readily converted to GD1b-disialoganglioside by bacterial and other mammalian neuraminidases without detergent or bile salts.
M.W. 2180.5 (based on stearoyl - 18:0).
Trisialoganglioside prepared from bovine gray matter. Readily converted to GD1b-disialoganglioside by bacterial and mammalian sialidases without detergent or bile salts. Inhibits mitogenesis in concanavalin A-stimulated murine T-cells. Lymphocyte marker in cancer patients.



Note: This product is not to be used for animal treatment, in vivo research or in any other contact procedure with livestock.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Inhibits Con A-stimulated mitogenesis in murine T cells
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Preparation Note

For use in aqueous systems, dissolve in a small amount of ethanol and then sonicate in buffer or medium, or dissolve the dried sample directly in aqueous buffers by sonication. Solutions can be filter sterilized.

Reconstitution

Following reconstitution, store in the refrigerator(4°C). Stock solutions are stable for up to 6 months at 4°C.

Other Notes

Hakomori, S. 1985. Cancer Res.45, 2405.
Momoi, T., et al. 1976. Biochim. Biophys. Acta441, 488.
Warren, L. 1959. J. Biol. Chem.234, 1971.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service