Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345724

Sigma-Aldrich

Ganglioside GM1, Ammonium Salt, Bovine Brain

Monosialoganglioside extracted from bovine brain.

Synonym(s):

Ganglioside GM1, Ammonium Salt, Bovine Brain, Galβ1,3GalNAcβ1,4(Neu5Acα2,3)Galβ1,4Glcβ1,1 Ceramide, NH₄

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C73H130N3O31 · NH4
CAS Number:
Molecular Weight:
1563.85
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

shipped in

ambient

storage temp.

−20°C

SMILES string

N([C@H]1[C@@H](O[C@@H]([C@@H]([C@@H]1O[C@@H]5O[C@@H]([C@@H]([C@@H]([C@H]5O)O)O)CO)O)CO)O[C@H]2[C@H](O[C@H]([C@@H]([C@H]2OCC(O)C(O)[C@@H]4O[C@](C[C@@H]([C@H]4NC(=O)C)O)(O)C(=O)O)O)O[C@@H]3[C@H](O[C@H]([C@@H]([C@H]3O)O)OCC(NC(=O)CCCCCCCCCCCCCCCCC)C(O)\C=C\C

InChI

1S/C73H131N3O31/c1-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-52(86)76-44(45(83)33-31-29-27-25-23-21-18-16-14-12-10-8-6-2)40-99-69-61(93)59(91)63(50(38-79)102-69)104-71-62(94)67(98-41-47(85)55(87)66-53(74-42(3)81)46(84)35-73(97,107-66)72(95)96)64(51(39-80)103-71)105-68-54(75-43(4)82)65(57(89)49(37-78)100-68)106-70-60(92)58(90)56(88)48(36-77)101-70/h31,33,44-51,53-71,77-80,83-85,87-94,97H,5-30,32,34-41H2,1-4H3,(H,74,81)(H,75,82)(H,76,86)(H,95,96)/b33-31+/t44?,45?,46-,47?,48+,49+,50+,51+,53+,54+,55?,56-,57-,58-,59+,60+,61+,62+,63+,64-,65+,66+,67+,68-,69+,70-,71-,73-/m0/s1

InChI key

CNLVNZJJSHZYAS-ALSOZVJRSA-N

Related Categories

General description

M.W. 1563.9 (based on stearoyl - 18:0).
Monosialoganglioside extracted from bovine brain. Useful as an antigen and receptor for cholera toxin, as a growth inhibition marker in fibroblasts, and as a marker for lymphoid subpopulations.
Monosialoganglioside extracted from bovine brain. Useful as an antigen and receptor for cholera toxin, as a growth inhibition marker in fibroblasts, and as a marker for lymphoid subpopulations. One mole yields 1 mole of sialic acid by thiobarbituric acid assay.



Note: This product is not to be used for animal treatment, in vivo research or in any other contact procedure with livestock.
One mole yields 1 mole of sialic acid by thiobarbituric acid assay.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Useful as an antigen and receptor for cholera toxin, as a growth inhibition marker in fibroblasts, and as a marker for lymphoid subpopulations.
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 3 weeks at 4°C or for up to 3 months at -20°C.

Other Notes

Gouy, H., et al. 1995. J. Immunol. 155, 5160.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service