Skip to Content
MilliporeSigma
All Photos(1)

Documents

345250

Millipore

N-Acetyllactosamine

A substrate for galactosidases, fucosyltransferases, and sialyltransferases. Useful as a lectin inhibitory sugar.

Synonym(s):

N-Acetyllactosamine, O-β-D-Galactopyranosyl-β1,4-2-acetamido-2-deoxy-D-glucose, Galβ1,4GlcNAc, LacNAc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25NO11
CAS Number:
Molecular Weight:
383.35
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C14H25NO11/c1-4(18)15-7-9(20)12(6(3-17)24-13(7)23)26-14-11(22)10(21)8(19)5(2-16)25-14/h5-14,16-17,19-23H,2-3H2,1H3,(H,15,18)/t5-,6-,7-,8+,9-,10+,11-,12-,13-,14+/m1/s1

InChI key

KFEUJDWYNGMDBV-LODBTCKLSA-N

General description

A substrate for galactosidases, fucosyltransferases, and sialyltransferases. Useful as a lectin inhibitory sugar.

Application


  • Mass Spectrometry Analysis of Glycopeptides Enriched by Anion Exchange-Mediated Methods Reveals PolyLacNAc-Extended N-Glycans in Integrins and Tetraspanins in Melanoma Cells.: This research demonstrates the application of mass spectrometry in identifying N-acetyllactosamine extensions on glycoproteins in melanoma cells, providing insights into cellular adhesion and signaling mechanisms (Čaval et al., 2024).


Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Analysis Note

Single spot by TLC

Other Notes

Gunnarsson, A., et al. 1984. Infect. Immun.45, 41.
Loomes, L.M., et al. 1984. Nature307, 560.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service