Skip to Content
MilliporeSigma

324717

Endoglycosidase H

Cleaves high-mannose N-glycans, suitable for Mass Spectrometry, from Streptomyces plicatus, recombinant, expressed in E. coli

Synonym(s):

Endoglycosidase H, Streptomyces plicatus, Recombinant, E. coli, Endo-β-N-acetylglucosaminidase H, Endo H

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
EC Number:
MDL number:
NACRES:
NA.54
UNSPSC Code:
12352202
Specific activity:
≥3 units/mL, ≥30 units/mg protein
Recombinant:
expressed in E. coli
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Endoglycosidase H, Streptomyces plicatus, Recombinant, E. coli, Endoglycosidase H, Streptomyces plicatus, Recombinant, E. coli cleaves between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide.

recombinant

expressed in E. coli

Quality Segment

conjugate

(N-linked)

form

liquid

specific activity

≥3 units/mL, ≥30 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

N-acetylglucosaminidase, α- and β-galactosidase, α-mannosidase, neuraminidases, proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Note: 1 mU = 1 milliunit.
Recombinant, streptomyces plicatus endoglycosidase H expressed in E. coli. Catalyzes the hydrolysis of asparagine-linked oligomannose and hybrid, but not complex oligosaccharides from glycoprotein. Endo H cleaves between the two N-acetylglucosamine residues in the diacetylchitobiose core of the oligosaccharide.

Physical form

In 50 mM NaCl, 20 mM Tris-HCl, 1 mM EDTA, pH 7.5.

Other Notes

One unit is defined as the amount of enzyme that will catalyze the release of N-linked oligosaccharides from 1.0 µmol denatured ribonuclease B per min at 37°C, pH 5.5. Cleavage is monitored by SDS-PAGE.
Rao, V., et al. 1995. Structure 3, 449.
Trimble, R.B., et al. 1987. Methods Enzymol. 138, 763.
Trumbly, R.J., et al. 1985. J. Biol. Chem.260, 5683.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library