Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

324696

Millipore

Elastase Substrate I, Colorimetric

A specific chromogenic substrate for human neutrophil elastase.

Synonym(s):

Elastase Substrate I, Colorimetric

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38N6O9
CAS Number:
Molecular Weight:
590.63
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)N[C@@H](C(C)C)C(=O)NC(=O)[C@H]2N(CCC2)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)CCC(=O)OC)C)C

InChI

1S/C27H38N6O9/c1-15(2)23(30-18-8-10-19(11-9-18)33(40)41)26(38)31-25(37)20-7-6-14-32(20)27(39)17(4)29-24(36)16(3)28-21(34)12-13-22(35)42-5/h8-11,15-17,20,23,30H,6-7,12-14H2,1-5H3,(H,28,34)(H,29,36)(H,31,37,38)/t16-,17-,20-,23-/m0/s1

InChI key

JWYKDZIEIWGSBN-OSAZLGQLSA-N

General description

A specific chromogenic substrate for human neutrophil elastase (Kcat/Km = 120,000 vs. 2700 for pancreatic elastase). Cleavage can be monitored at ~410 nm. Sold on the basis of substrate content.
Specific chromogenic substrate for human neutrophil elastase (kcat/Km = 120,000 versus pancreatic elastase: kcat/Km = 2,700). Cleavage can be monitored at 410 nm using ε = 8,800.

Biochem/physiol Actions

Cell permeable: no
Kcat/Km = 120,000 vs. 2700 for pancreatic elastase
Primary Target
Chromogenic substrate for human neutrophil elastase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

MeOSuc-Ala-Ala-Pro-Val-pNA

Physical form

Lyophilized from a solution containing 90 mg D-mannitol and 10 mg substrate.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Barrett, A.J., et al. 1989. Biochim. J.260, 259.
Nakajima, K., et al. 1979. J. Biol. Chem.254, 4027.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Luan et al.
Toxins, 11(12) (2019-12-11)
Elastase is a globular glycoprotein and belongs to the chymotrypsin family. It is involved in several inflammatory cascades on the basis of cleaving the important connective tissue protein elastin, and is strictly regulated to a balance by several endogenous inhibitors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service