Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

259580

Sigma-Aldrich

2-Deoxy-D-galactose

Hexose analog that can be incorporated into rat gangliosides GM2 and GD3 in positions normally occupied by galactose.

Synonym(s):

2-Deoxy-D-galactose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C6H12O5/c7-2-1-4(9)6(11)5(10)3-8/h2,4-6,8-11H,1,3H2/t4-,5-,6-/m1/s1

InChI key

VRYALKFFQXWPIH-HSUXUTPPSA-N

General description

Hexose analog that can be incorporated into rat gangliosides GM2 and GD3 in positions normally occupied by galactose. 2-Deoxy-D-galactose has been suggested to act as an inhibitor of fucosylation. It has also been used for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).
Hexose analog that can be incorporated into rat gangliosides GM2 and GD3 in positions normally occupied by galactose. 2-Deoxy-D-galactose has been suggested to act as an inhibitor of fucosylation. It has also been used for competitive elution of Anadarin P lectin (a galactosyl-binding lectin from blood clam).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up 6 months at -20°C.

Other Notes

Lorenzini, C.G., et al. 1997. Neurobiol. Learn. Mem. 68, 317.
Kohla, G., et al. 1993. FEBS Lett. 327, 63.
Dam, T.K. 1992. Mol. Cell. Biochem. 117, 1.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service