Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239900

Sigma-Aldrich

Cypermethrin

Synthetic type II pyrethroid. Potent inhibitor of calcineurin (protein phosphatase 2B; IC₅₀ = 40 pM). May also have varied effects on ion channels.

Synonym(s):

Cypermethrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19Cl2NO3
CAS Number:
Molecular Weight:
416.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

description

Merck USA index - 14, 2767

Quality Level

Assay

≥95% (HPLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: 15 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

ClC(=CC1C(C1C(=O)OC(c2cc(ccc2)Oc3ccccc3)C#N)(C)C)Cl

InChI

1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3

InChI key

KAATUXNTWXVJKI-UHFFFAOYSA-N

General description

Synthetic type II pyrethroid. Potent inhibitor of calcineurin (protein phosphatase 2B; IC50 = 40 pM). May also have varied effects on ion channels.
Type II pyrethroid insecticide. Potent inhibitor of calcineurin (protein phosphatase 2B, IC50 = 40 pM). May also have varied effects on ion channels.

Biochem/physiol Actions

Primary Target
calcineurin
Product does not compete with ATP.
Target IC50: 40 pM against calcineurin (protein phosphatase 2B

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Richter, A., et al. 1995. Biochem. Pharmacol.49, 367.
Wang, J.H., and Stelzer, A. 1994. NeuroReport 5, 2377.
Enan, E., and Matsumura, F. 1992. Biochem. Pharmacol.43, 1777.
Narahasi, T. 1992. Trends Pharmacol. Sci.13, 236.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Nervous system, Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service