Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

219365

Sigma-Aldrich

Coagulation Factor V, Human Plasma

Coagulation Factor V, Human Plasma, CAS 9001-24-5, is a native coagulation factor V that is cleaved by thrombin to yield activated Factor Va that is 50-fold greater activity than its precursor form.

Synonym(s):

Coagulation Factor V, Human Plasma, Proaccelerin, Labile Factor, Ac-globulin, Ac-g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

description

Merck USA index - 14, 3919

Assay

≥95% (SDS-PAGE)

form

liquid

specific activity

≥20 units/mg protein (clotting assay)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

wet ice

storage temp.

−20°C

General description

Native coagulation factor V from human plasma. Thrombin cleaves Factor V to yield activated Factor Va, which exhibits 50-fold greater activity than the precursor molecule. Factor Va, Ca2+, and membrane phospholipids are cofactors which form a "prothrombinase" complex with Factor Xa, enabling its proteolytic activation of prothrombin to thrombin.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50% glycerol/H₂O (v/v).

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Do not store below -20°C.

Analysis Note

Clotting activity is determined by comparison to a standard curve produced using normal pooled human plasma

Other Notes

Walker, F.J. 1992. J. Biol. Chem.267, 19896.
Monkovic, D.D., and Tracy, P.B. 1990. J. Biol. Chem.265, 17132.
Krishnaswamy, S., et al. 1989. J. Biol. Chem.264, 3160.
Lee, C.D., and Mann, K.G. 1989. Blood73, 185.
Krieg, U.C., et al. 1987. Biochemistry26, 103.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service