Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

218728

Sigma-Aldrich

Caspase-9 Inhibitor III

The Caspase-9 Inhibitor III, also referenced under CAS 403848-57-7, controls the biological activity of Caspase-9. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase-9 Inhibitor III, Ac-LEHD-CMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H35ClN6O9
CAS Number:
Molecular Weight:
587.02
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent and irreversible inhibitor of caspase-9 (ICE-LAP6, Mch6). Reported to reduce myocardial infarct size during reperfusion (~70 nM).
A potent, irreversible inhibitor of caspase-9. Reported to reduce myocardial infarct size during reperfusion (~70 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
caspase-9
Product does not compete with ATP.
Reversible: no
Target IC50: 70 nM against caspase-9

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Leu-Glu-His-Asp-CMK

Other Notes

Mocanu, M.M., et al. 2000. Br. J. Pharmacol.130, 197.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hazel Stewart et al.
iScience, 26(11), 108080-108080 (2023-10-20)
The SARS-CoV-2 genome encodes a multitude of accessory proteins. Using comparative genomic approaches, an additional accessory protein, ORF3c, has been predicted to be encoded within the ORF3a sgmRNA. Expression of ORF3c during infection has been confirmed independently by ribosome profiling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service