Skip to Content
MilliporeSigma
All Photos(1)

Documents

2120-OP

Millipore

Agarose

OmniPur® Grade

Synonym(s):

OmniPur® Agarose, 3,6-Anhydro-α-L-galacto-β-D-galactan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41105317
NACRES:
NA.25

Quality Level

form

fine powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

storage temp.

room temp

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

General description

Specifications
DNAse: none detected
RNAse: none detected
Protease: none detected
Sulfate (SO4): ≤0.15%
EEO (-mr): ≤0.1
Gelling range (1%): 36-39 °C
Gel strength (1%): 1200 g/cm2 min
Melting point (1%): 87-89 °C

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service