Select a Size
About This Item
Quality Segment
assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
mp
60-90 °C
bulk density
700 kg/m3
shipped in
ambient
storage temp.
−20°C
SMILES string
O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O
InChI
1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1
InChI key
MJQHZNBUODTQTK-WKGBVCLCSA-N
Application
- Porous Agarose Layered Magnetic Graphene Oxide Nanocomposite for Virus RNA Monitoring in Wastewater.: Introduces a composite material incorporating agarose for enhanced viral RNA capture and detection in environmental samples, highlighting agarose′s utility in public health monitoring (He B et al., 2024).
Analysis Note
Appearance (description): crystalline powder
Identity (IR-spectrum): passes test
Electroendoosmosis: 0.05 - 0.13
Gel point: 34 - 38 °C
Sulphur (as SO₄): ≤ 0.3 %
Sulfated ash (800 °C): ≤ 1.0 %
Loss on drying (105 °C, 2 h): ≤ 10.0 %
Legal Information
Disclaimer
Still not finding the right product?
Explore all of our products under Agarose
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1168020025 | 04022536168173 |
| 1168020250 | 04022536168180 |