Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20845-M

Sigma-Aldrich

IHC Select 20X TBS Rinse Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.47

form

liquid

Quality Level

manufacturer/tradename

Chemicon®
IHC Select

technique(s)

immunohistochemistry: suitable (paraffin)

Application

For use as a rinse buffer in Immunohistochemical staining procedures after dilution (see preparation below). Use prepared 1X Rinse Buffer at room temperature.



For best results use this reagent in conjunction with other IHC Select detection reagents. A complete list of reagents is available at www.chemicon.com.

Physical form

Liquid TBS containing 0.4% Tween 20 and 0.1% Kathon as a preservative. Preparation: Before use, dilute this 20X Rinse Buffer to 1X with distilled or deionized water. For example, to make 1 liter of 1X Rinse Buffer, add 50 mL of 20X Rinse Buffer to 950 mL of water and mix thoroughly. For improved results it is recommended (but not required) that Tween 20 be added to the 1X Rinse Buffer at a final concentration of 0.1% (1.0 mL of Tween 20 for each 1 liter of 1X Rinse Buffer) and mix thoroughly.

Store prepared 1X Rinse Buffer at 2-8ºC. Warm to room temperature before use. Discard 1X Rinse Buffer if it becomes cloudy or contaminated.

Storage and Stability

When stored at 2-8°C, the 20X Rinse Buffer is stable up to the expiration date printed on the label. Do not freeze or expose to elevated temperatures.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service