Skip to Content
MilliporeSigma

Skip To

935B

TBS IHC Wash Buffer + TWEEN® 20

20X TBS, 1% Tween® 20

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

[For In Vitro Diagnostic Use in Select Regions]

form

buffered aqueous solution

packaging

pkg of 1 L 20x (935B-09), pkg of 200 mL 20x (935B-06)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Tris Buffered Immunohistochemistry wash buffer plus TWEEN 20 is a 20X concentrated solution that is employed to rinse reagents off slides and to provide a medium for short-term storage of immunohistochemistry specimens in between application of reagents.

Biochem/physiol Actions

Tween 20 helps in the enhancement of intestinal movement of doxorubicin in vitro but not in vivo.

Physical form

Solution containing TBS buffer and TWEEN 20

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
934BEF5013524653
description

20X TBS, 1% Tween® 20

description

20X PBS, 1% Tween® 20

description

EF5 antibody dilution buffer specifically formulated for use with the EF5 Hypoxia Detection Kits and anti-EF5 monoclonal antibodies.

description

Dissolve one tablet in 1 liter of deionized water to yield 140 mM NaCl, 3 mM KCl, 0.05% TWEEN 20 Detergent, 10 mM phosphate buffer, pH 7.4 at 25°C. Note: 1 each = 10 tablets.

technique(s)

immunohistochemistry: suitable

technique(s)

immunohistochemistry: suitable

technique(s)

-

technique(s)

-

Quality Level

500, 600

Quality Level

500, 600

Quality Level

100

Quality Level

100

form

buffered aqueous solution

form

buffered aqueous solution

form

liquid

form

tablet

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

15-25°C

description

[For In Vitro Diagnostic Use in Select Regions]

description

[For In Vitro Diagnostic Use in Select Regions]

description

-

description

-

packaging

pkg of 1 L 20x (935B-09), pkg of 200 mL 20x (935B-06)

packaging

pkg of 1 L 20x (934B-09), pkg of 200 mL 20x (934B-06)

packaging

-

packaging

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Is there a recommended or preferable wash buffer (TBS or PBS) for a breast panel in Autostainer? Moreover, please confirm how long one can wait once the dilution of the concentrated product is done? Both TBS and PBS in this case.

    1 answer
    1. Phosphate Buffer Saline (PBS) and Tris Buffer Saline (TBS) are concentrated wash solutions designed for rinsing reagents from tissue slides and preserving tissue sections between reagent applications. PBS is recommended with any detection system, except those containing alkaline phosphatase. Conversely, TBS is recommended when alkaline phosphatase is present to avoid excessive phosphate accumulation and consequent reduction in the rate of reaction. Adding Tween® 20 to both PBS and TBS may help reduce background and produce a cleaner immunohistochemical stain overall. The most significant factor is the intention for use to visualize the stain, with what chromogen and detection kit. For HRP or Alk Phos on the breast panel, TBS can be used for both. For diluent, as long as products are stored properly (2 to 8C), the diluent will help stabilize diluted antibodies, and the same applies for use with HRP or Alk Phos detection for TBS to be run on either, while PBS works best with HRP. This allows for a longer expiration date and aligns with the antibody expiration date.

      Helpful?

Reviews

No rating value

Active Filters