Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

203297-M

Millipore

Bisindolylmaleimide IV

A potent, cell permeable, and selective inhibitor of protein kinase C (IC₅₀ = 87 nM).

Synonym(s):

Bisindolylmaleimide IV, Arcyriarubin A, 2,3- bis(1H-Indol-3-yl)maleimide, Ro 31-6233

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13N3O2
CAS Number:
Molecular Weight:
327.34
MDL number:
UNSPSC Code:
12352111

Quality Level

Assay

≥98% (TLC)

form

powder
solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

dark orange

solubility

methanol: 10 mg/mL

storage temp.

2-8°C

SMILES string

N1C(=O)C(=C(C1=O)c4c5c([nH]c4)cccc5)c2c3c([nH]c2)cccc3

InChI

1S/C20H13N3O2/c24-19-17(13-9-21-15-7-3-1-5-11(13)15)18(20(25)23-19)14-10-22-16-8-4-2-6-12(14)16/h1-10,21-22H,(H,23,24,25)

InChI key

DQYBRTASHMYDJG-UHFFFAOYSA-N

General description

A cell-permeable potent and selective inhibitor of protein kinase C (IC50 = 87 nM) and protein kinase A (IC50 = 2.7 µM).
A potent, cell permeable, and selective inhibitor of protein kinase C (IC50 = 87 nM). Inhibits protein kinase A at much higher concentrations (IC50 = 2.7 µM).

Biochem/physiol Actions

Primary Target
PKC
Target IC50: 87 nM against protein kinase C

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Fabre, S., et al. 1993. Bioorg. Med. Chem.1, 193.
Davis, P.D., et al. 1992. J. Med. Chem.35, 177.
Toullec, D., et al. 1991. J. Biol. Chem. 266, 15771.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service