Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B4056

Sigma-Aldrich

Bisindolylmaleimide XI hydrochloride

≥98% (TLC), solid

Synonym(s):

Ro 32-0432

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28N4O2 · HCl
Molecular Weight:
489.01
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

solid

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

Cl.CN(C)CC1CCn2c(C1)cc3ccc(cc23)C4=C(C(=O)NC4=O)c5cn(C)c6ccccc56

InChI

1S/C28H28N4O2.ClH/c1-30(2)15-17-10-11-32-20(12-17)13-18-8-9-19(14-24(18)32)25-26(28(34)29-27(25)33)22-16-31(3)23-7-5-4-6-21(22)23;/h4-9,13-14,16-17H,10-12,15H2,1-3H3,(H,29,33,34);1H

InChI key

WMVQGKBSNAIMHI-UHFFFAOYSA-N

Biochem/physiol Actions

Selective inhibitor of protein kinase C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillaume Robert et al.
PloS one, 4(11), e7889-e7889 (2009-11-20)
CML is an hematopoietic stem cell disease characterized by the t(9;22) (q34;q11) translocation encoding the oncoprotein p210BCR-ABL. The effect of acadesine (AICAR, 5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside) a compound with known antileukemic effect on B cell chronic lymphoblastic leukemia (B-CLL) was investigated in different
Caline S Karam et al.
Journal of chemical neuroanatomy, 83-84, 91-98 (2017-06-03)
The dopamine transporter (DAT), which mediates the inactivation of released dopamine through its reuptake, is the primary molecular target for the actions of psychostimulants. An increasing number of studies support an essential role for phosphorylation of serines (Ser) in the
M Signore et al.
Cell death & disease, 5, e1223-e1223 (2014-05-09)
Glioblastoma (GBM) is the most common and deadly adult brain tumor. Despite aggressive surgery, radiation, and chemotherapy, the life expectancy of patients diagnosed with GBM is ∼14 months. The extremely aggressive nature of GBM results from glioblastoma stem-like cells (GSCs)
Florence P Varodayan et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(17), 4593-4603 (2017-04-02)
L-type voltage-gated calcium channels (LTCCs) are implicated in several psychiatric disorders that are comorbid with alcoholism and involve amygdala dysfunction. Within the amygdala, the central nucleus (CeA) is critical in acute alcohol's reinforcing actions, and its dysregulation in human alcoholics
S Khom et al.
Science advances, 6(12), eaaz1050-eaaz1050 (2020-03-25)
Behavioral and clinical studies suggest a critical role of substance P (SP)/neurokinin-1 receptor (NK-1R) signaling in alcohol dependence. Here, we examined regulation of GABA transmission in the medial subdivision of the central amygdala (CeM) by the SP/NK-1R system, and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service