Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

200484-M

Sigma-Aldrich

Bestatin

Binds to cell surfaces and inhibits surface aminopeptidases, notably aminopeptidase B and leucine aminopeptidase.

Synonym(s):

Bestatin, [(2S,3R)-3-Amino-2-hydroxy-4-phenylbutanoyl]-Leu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2O4
CAS Number:
Molecular Weight:
308.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

description

Merck USA index - 14, 9842

Assay

>99% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 2 mg/mL
methanol: 5 mg/mL

storage temp.

2-8°C

SMILES string

[N+H3][C@@H]([C@H](O)C(=O)N[C@H](CC(C)C)C(=O)[O-])Cc1ccccc1

InChI

1S/C16H24N2O4/c1-10(2)8-13(16(21)22)18-15(20)14(19)12(17)9-11-6-4-3-5-7-11/h3-7,10,12-14,19H,8-9,17H2,1-2H3,(H,18,20)(H,21,22)/t12-,13-,14+/m1/s1

InChI key

VGGGPCQERPFHOB-MCIONIFRSA-N

General description

Binds to cell surfaces and inhibits surface aminopeptidases, notably aminopeptidase B and leucine aminopeptidase. Also acts as a potent aminopeptidase N inhibitor. Activates macrophages and T lymphocytes. Has antitumor properties. Also available as a 50 mM solution in water (Cat. No. 200488).

Biochem/physiol Actions

Primary Target
amino peptidase b

Warning

Toxicity: Standard Handling (A)

Preparation Note

Unstable in aqueous solution; dilute just prior to use.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO and methanol stock solutions are stable for up to 1 month at -20°C.

Other Notes

Inoue, T., et al. 1994. J. Clin. Endocrinol. Metab. 79, 171.
Taylor, A., et al. 1993. Biochemistry32, 784.
Ishida, J.I., et al. 1984. J. Chromatogr.305, 381.
Umezawa, H. 1982. Annu. Rev. Microbiol.36, 75.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service