Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PEPS-RO

Roche

Pepstatin

from Streptomyces species

Synonym(s):

Pepstatin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H63N5O9
CAS Number:
Molecular Weight:
685.89
Enzyme Commission number:
Beilstein:
2201362
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

biological source

synthetic

Quality Level

form

lyophilized

packaging

pkg of 10 mg (11359053001)
pkg of 2 mg (10253286001)
pkg of 50 mg (11524488001)

manufacturer/tradename

Roche

technique(s)

affinity chromatography: suitable
protein purification: suitable

mp

233 °C (dec.) (lit.)

solubility

ethanol: 1 mg/mL
methanol: 1 mg/mL
acetic acid: soluble 300 μg/mL (6N)
pyridine: soluble
water: insoluble

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](C)NC(=O)C[C@H](O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)[C@@H](O)CC(O)=O

InChI

1S/C34H63N5O9/c1-17(2)12-23(37-33(47)31(21(9)10)39-34(48)30(20(7)8)38-27(42)14-19(5)6)25(40)15-28(43)35-22(11)32(46)36-24(13-18(3)4)26(41)16-29(44)45/h17-26,30-31,40-41H,12-16H2,1-11H3,(H,35,43)(H,36,46)(H,37,47)(H,38,42)(H,39,48)(H,44,45)/t22-,23-,24-,25-,26-,30-,31-/m0/s1

InChI key

FAXGPCHRFPCXOO-LXTPJMTPSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Isovaleryl-L-val-L-val-statyl-L-alanyl-statin (Isovaleryl-L-val-L-val-4-(S)-amino-3-(S)-hydroxy-6-methyl-heptanoyl-2-ala-4-(S)-amino-3-(S)-hydroxy-6-methyl-heptanoic acid)
Pepstatin is a low molecular weight, highly specific inhibitor of acid proteases (in particular aspartate proteases).

Specificity

Pepstatin potently inhibits the HIV protease and other aspartic proteases, such as pepsin, renin, cathepsin D, chymosin, protease B from Aspergillus niger, and acid proteases of microbial origin.

Application

Pepstatin is used for the protection of proteins and enzymes during isolation and purification procedures. It facilitates the investigation of enzyme mechanisms and biological function, and the classification of proteases. Pepstatin is applied in affinity chromatography, structural and pharmacological research, in HIV research (inhibition of retroviral-coded proteases, e.g., HIV-1), and cancer research (as an immunostimulant and anti-tumor drug).

Biochem/physiol Actions

Pepstatin A is an inhibitor of acid proteases (aspartyl peptidases). It forms a 1:1 complex with proteases such as pepsin, renin, cathepsin D, bovine chymosin, and protease B (Aspergillus niger). The inhibitor is highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases. Solubilized γ-secretase and retroviral protease are also inhibited by Pepstatin A. It has been used to characterize proteases from several sources.

Quality

Purity tested by HPLC.

Preparation Note

Working concentration: 0.7 μg/ml (1 μM)
Working solution: Recommended solvent is methanol (1 mg/ml), ethanol, DMSO or glacial acetic acid.
Pepstatin is insoluble in pure water, but it is possible to produce an aqueous solutions with methanol (1.5–2.0 mg/ml), ethanol (1 mg/ml if allowed to sit overnight) or DMSO. Pepstatin is also soluble in acetic acid (6 N, 300 μg/ml) and pyridine. Solubility in NaCl solution is not known. Sodium or magnesium salts of pepstatin are water-soluble.
Stock solution: 1.4 mg pepstatin/mI (2 mM), in methanol
Storage conditions (working solution): -15 to -25 °C
Pepstatin can be dissolved at 1 mg/ml in 10% (v/v) acetic acid in methanol (9:1 methanol:acetic acid). The inclusion of acetic acid may be necessary to dissolve this peptide in methanol or DMSO. It has been dissolved at 1 to 2 mg/ml in ethanol, but heat may be required for complete dissolution. Solutions of Pepstatin A can be heated as high as 60 °C without any decomposition of the peptide.
Stock solutions at 1 mg/ml are stable at least a week at 4 °C. A 1 mM solution in methanol or DMSO is stable for months at -20 °C. If solutions become darker yellow, the reagent is hydrolyzing.
A working concentration of 1 μM is stable for at least one day at room temperature.

Reconstitution

Soluble in methanol to approximately 1 mg/ml. Also soluble to 1 mg/ml in ethanol if allowed to sit overnight, and to 300 μg/ml in 6 N acetic acid. Stable for at least one week at 2 to 8 °C, or one month when stored in aliquots at -15 to -25 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service