Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

200274

Sigma-Aldrich

Nα-Benzyloxycarbonyl-L-lysine Thiobenzyl Ester, Hydrochloride

Highly sensitive substrate for trypsin-like enzymes and granule-associated serine esterase.

Synonym(s):

Nα-Benzyloxycarbonyl-L-lysine Thiobenzyl Ester, Hydrochloride, BLT, HCl

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.77

Quality Level

Assay

>95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white to off-white

solubility

water: 10 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

S(Cc2ccccc2)C(=O)[C@@H](NC(=O)OCc1ccccc1)CCCCN.[Cl-].[H+]

InChI

1S/C21H26N2O3S.ClH/c22-14-8-7-13-19(20(24)27-16-18-11-5-2-6-12-18)23-21(25)26-15-17-9-3-1-4-10-17;/h1-6,9-12,19H,7-8,13-16,22H2,(H,23,25);1H/t19-;/m0./s1

InChI key

PNDSXGXPDJRQAV-FYZYNONXSA-N

General description

Highly sensitive substrate for trypsin-like enzymes and granule-associated serine esterases.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Substrate for trypsin-like enzymes and granule-associated serine esterase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Green, G.D.J. and Shaw, W. 1979. Anal. Biochem. 93, 223.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service