Skip to Content
MilliporeSigma

N8010

4-Nitrophenyl 4-guanidinobenzoate hydrochloride

protease substrate, ≥95.0% (TLC), powder

Synonym(s):

4-Guanidinobenzoic acid 4-nitrophenylester hydrochloride, NPGB, pNPGB

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H12N4O4 · HCl
CAS Number:
Molecular Weight:
336.73
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
242-831-3
MDL number:
Beilstein/REAXYS Number:
8024554
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

4-Nitrophenyl 4-guanidinobenzoate hydrochloride, protease inhibitor and substrate

Quality Segment

assay

≥95.0% (TLC)

form

powder

solubility

formic acid: soluble 49.00-51.00 mg/mL

storage temp.

−20°C

SMILES string

Cl[H].NC(=N)Nc1ccc(cc1)C(=O)Oc2ccc(cc2)[N+]([O-])=O

InChI

1S/C14H12N4O4.ClH/c15-14(16)17-10-3-1-9(2-4-10)13(19)22-12-7-5-11(6-8-12)18(20)21;/h1-8H,(H4,15,16,17);1H

InChI key

PKSBDZOBYIKNGY-UHFFFAOYSA-N

General description

4-Nitrophenyl 4-guanidinobenzoate hydrochloride (pNPGB) is a trypsin substrate. It is more water-soluble than 4-methylumbelliferyl 4-guanidinobenzoate and is preferred in spectrophotometric assay.

Application

4-Nitrophenyl 4-guanidinobenzoate hydrochloride has been used:
  • as a substrate for trypsin for active site titration experiments
  • for pre-treating of mosquito eggs in the interplasmid transposition assay
  • as a component of isotonic buffer to moisten filter paper for mosquito embryo collection

Biochem/physiol Actions

4-Nitrophenyl 4-guanidinobenzoate hydrochloride (pNPGB) inhibits phenoloxidase enzyme and delays embryo chorionic membrane hardening.


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library