Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

176905

Sigma-Aldrich

Anthrax Protective Antigen

from Recombinant, >90% (SDS-PAGE), lyophilized, anthrax protective antigen, Calbiochem®

Synonym(s):

Anthrax PA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Anthrax Protective Antigen, Bacillus anthracis, Recombinant, B. anthracis, Anthrax Protective Antigen, Bacillus anthracis, Recombinant, is the protective antigen that mediates the entry of anthrax lethal factor and edema factor into the target cell.

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Recombinant, Bacillus anthracis anthrax protective antigen expressed in a special strain of B. anthracis. One of three protein components of anthrax toxin produced by the pathogenic bacterium, Bacillus anthracis. Central moiety that mediates entry of lethal factor (LF) and edema factor (EF) into the target cell. PA83 binds to the cell surface via a type I membrane receptor with a von Willebrand factor A domain, known as anthrax toxin receptor. PA83 is proteolytically activated in vivo by a furin-like protease to produce a 63 kDa protein (PA63). PA63 self-associates to form a heptameric, cation-selective, voltage-gated, membrane channel that subsequently binds to LF and/or EF. Cleavage of the 20 kDa fragment from PA83 is necessary for binding of protective antigen to LF and EF.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Type I membrane protein with a von Willebrand factor A domain called anthrax toxin receptor
Product does not compete with ATP.
Reversible: no

Packaging

yes

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 50 mM NaCl, 5 mM HEPES, pH 7.5.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 6 months at -20°C.
Reconstitute in 100 µl sterile, distilled H₂O to yield a final concentration of 1 mg/ml.

Analysis Note

Single major band by SDS-PAGE. Several smaller minor components are apparent that may represent cleavage fragments of the PA.

Other Notes

Mogridge, J., et al. 2002. Biochemistry41, 1079.
Nassi, S., et al. 2002. Biochemistry41, 1445.
Ahuja, N., et al. 2001. Biochem. Biophys. Res. Commun.286, 6.
Bradley, K.A., et al. 2001. Nature414, 225.
Brossier, F., and Mock, M. 2001. Toxicon39, 1747.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service