Skip to Content
MilliporeSigma
All Photos(2)

Documents

17-10451

Sigma-Aldrich

CpGenome Direct Prep Bisulfite Modification Kit (50 Reactions)

The CpGenome Direct Prep Bisulfite Modification Kit allows bisulfite conversion directly from a variety of starting materials, including cultured cells, blood, fresh tissue & fixed tissue samples.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

manufacturer/tradename

CpGenome

application(s)

genomic analysis

Related Categories

General description

Methylation of cytosines located 5′ to guanosine is known to have a profound effect on the expression of many eukaryotic genes. In normal cells methylation occurs predominantly in CG-poor regions, while CG-rich areas, called CpG-islands remain unmethylated. The exceptions are the extensive methylation of CpG islands associated with transcriptional inactivation of regulatory regions of imprinted genes and genes on the inactive X-chromosome of females. Aberrant methylation of normally unmethylated CpG islands has been documented as a relatively frequent event in immortalized and transformed cells and has been associated with transcriptional inactivation of defined tumor suppressor genes in human cancers. Hundreds of CpG islands are now known to exhibit the characteristic of hypermethylation in tumors resulting in identification of candidate genes that can be interrogated to determine extent of tumor specific transformation.
Unlike other bisulfite modifications approaches that start with isolated genomic DNA, the CpGenome Direct Prep Bisulfite Modification Kit allows simple and reliable bisulfite conversion of DNA directly from a variety of starting materials, including cultured cells, blood, fresh tissue and fixed tissue samples.

Key Features
  • Bisulfite conversion directly from cells, tissues, blood and FFPE samples without DNA purification
  • Sensitive bisulfite-conversion from as few as 10 cells or as low as 50 pg of input DNA
  • Fast and simple, streamlined protocol for one-step bisulfite conversion
  • In-column desulfonation allows recovery of DNA without additional precipitation steps for more consistent results
  • Suitable for downstream analysis by methlylation specific PCR, restriction digestion, sequencing, microarray hybridization, etc.

Application

Research Category
Epigenetics & Nuclear Function

Components

2X Extraction Buffer

Modification Reagent

Conversion Buffer

Resuspension Buffer

Equilibration Buffer

Binding Buffer

Wash Buffer I

Wash Buffer II

Elution Buffer

Proteinase K

Proteinase K Storage Buffer

Desalting Columns

Collection Tubes

Linkage

Replaces: Replaces S7820 (CpGenome Universal DNA Modification Kit).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic and epigenetic alterations in normal and sensitive COPD-diseased human bronchial epithelial cells repeatedly exposed to air pollution-derived PM2.5.
Leclercq, B; Platel, A; Antherieu, S; Alleman, LY; Hardy, EM; Perdrix, E; Grova et al.
Environmental Pollution (Barking, Essex : 1987) null
Promoter hypermethylation of the cysteine protease RECK may cause metastasis of osteosarcoma.
Wang, L; Ge, J; Ma, T; Zheng, Y; Lv, S; Li, Y; Liu, S
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine null
Effects of 5-aza-2'deoxycytidine on RECK gene expression and tumor invasion in salivary adenoid cystic carcinoma.
Zhou, XQ; Huang, SY; Zhang, DS; Zhang, SZ; Li, WG; Chen, ZW; Wu, HW
Brazilian journal of medical and biological research null
Michael W Lewis et al.
Human molecular genetics, 28(2), 220-229 (2018-09-28)
Angelman syndrome, Prader-Will syndrome and Dup15q syndrome map to a cluster of imprinted genes located at 15q11-q13. Imprinting at this domain is regulated by an imprinting control region consisting of two distinct elements, the Angelman syndrome imprinting center (AS-IC) and
Li-Min Gao et al.
The American journal of surgical pathology, 40(6), 836-846 (2016-03-15)
Aggressive natural killer cell leukemia (ANKL) is a rare disease with an extremely aggressive clinical course. The etiology of ANKL is unclear with few genetic/epigenetic aberrations described to date. Moreover, misdiagnosis of ANKL is a frequent problem. Clinicopathologic characteristics of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service