Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.52250

Supelco

Chromolith® WP 300 Epoxy (monolithic) HPLC Columns

L × I.D. 100 mm x 4.6 mm HPLC Column

Synonym(s):

Chromolith® WP 300 Epoxy 100-4.6 HPLC column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

Product Name

Chromolith® WP 300 Epoxy, L × I.D. 100 mm x 4.6 mm HPLC column

material

PEEK column

Quality Level

product line

Chromolith®
Chromolith®

feature

endcapped: no
endcapped: no

packaging

1 ea of

extent of labeling

5-6% Carbon loading

parameter

2 - 60 °C temp. range
2-60 °C temp. range
200 bar pressure
200 bar pressure

technique(s)

HPLC: suitable
HPLC: suitable
LC/MS: suitable

column L × I.D.

10 cm × 4.6 mm

matrix

monolithic silica
monolithic silica

matrix active group

Epoxy bonding phase
Epoxy bonding phase

pore size

2 μm macroporosity
2 μm macroporosity
30 nm mesoporosity
30 nm mesoporosity

operating pH range

1.5-7.5
1.5-7.5

storage temp.

no temp limit

Looking for similar products? Visit Product Comparison Guide

General description

Chromolith® WP 300 Epoxy columns are specially designed for the user-specific immobilization of ligands and their later application in HPLC. The unique bimodal pore structure of silica monoliths allows efficient coupling independent of molecule size. The wider mesopores also enable the use of proteins and antibodies as both ligand immobilized on the column, and later analyte separated by an immobilized column.
Potential applications: attach trypsin to obtain HPLC column-protein digestion reactor; attach protein and measure other protein interaction with the attached one; attach any chiral selector to obtain a chiral column; attach any affinity ligand to obtain custom made affinity column, etc.

Chromolith® WP 300 Epoxy 100-4.6 columns are made of monolithic silica with a characteristic bimodal pore structure.

Key Benefits:
  • Easy to perform immobilization of ligand opens up limitless customization options
  • Monolithic format allows for high flow rates at low backpressure, enabling high throughput


Learn more:
Brochure: Race through your Separations on Any System. Chromolith® Monolithic Silica HPLC Columns

Chromolith® WP 300 Epoxy is available in 2 mm I.D. providing improved efficiency, sensitivity, and enhanced MS-compatibility.
Flyer: Create Your Own Column Chemistry - Chromolith® WP 300 Epoxy 2 mm I.D. HPLC Columns

All Supelco HPLC Columns, including monolithic Chromolith® columns, are fully compatible to all HPLC and UHPLC Instruments.
Supelco HPLC Products: The Best Choice for Any LC Instrument

Legal Information

Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

HPLC separates and identifies large biomolecules like proteins and peptides by pumping the sample through a sorbent-packed column.

HPLC separates and identifies large biomolecules like proteins and peptides by pumping the sample through a sorbent-packed column.

HPLC separates and identifies large biomolecules like proteins and peptides by pumping the sample through a sorbent-packed column.

HPLC separates and identifies large biomolecules like proteins and peptides by pumping the sample through a sorbent-packed column.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service