Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14-689-M

Sigma-Aldrich

Casein Kinase 2α2 Protein, active, 10 µg

Active, N-terminal 6His-tagged, recombinant, full length, human Casein Kinase 2α2, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

human

Quality Level

100
300

recombinant

expressed in Sf21 cells

mol wt

Mw 45.8 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal 6His-tagged, recombinant, full length, human Casein Kinase 2α2

Biochem/physiol Actions

Protein Target: CK2α2

Quality

routinely evaluated by phosphorylation of casein kinase 2 substrate peptide

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein kinase CK2 as regulator of cell survival: implications for cancer therapy.
Unger, G M, et al.
Current Cancer Drug Targets, 4, 77-84 (2004)
The genes encoding human protein kinase CK2 and their functional links.
Pyerin, Walter and Ackermann, Karin
Progress in Nucleic Acid Research and Molecular Biology, 74, 239-273 (2003)
Supervision of multiple signaling protein kinases by the CK2-Cdc37 couple, a possible novel cancer therapeutic target.
Miyata, Yoshihiko and Nishida, Eisuke
Annals of the New York Academy of Sciences, 1030, 150-157 (2004)
Order or chaos? An evaluation of the regulation of protein kinase CK2.
Olsten, Mary Ellen K and Litchfield, David W
Biochemistry and Cell Biology = Biochimie Et Biologie Cellulaire, 82, 681-693 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service