Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

07-496-I

Sigma-Aldrich

Anti-Myeloperoxidase Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Myeloperoxidase, myeloperoxidase heavy chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MPO(4353)

General description

Myeloperoxidase (EC 1.11.2.2; UniProt P05164) is encoded by the MPO gene (Gene ID 4353) in human. Myeloperoxidase/MPO is a heme protein synthesized during myeloid differentiation that constitutes the major component of neutrophil azurophilic granules. Produced as a 745 a.a. single chain precursor with a signal peptide sequence (a.a. 1-48), myeloperoxidase is subsequently cleaved into a light chain (a.a.165-278) and a heavy chain (a.a. 279-745). The mature myeloperoxidase is a tetramer composed of 2 light chains and 2 heavy chains. MPO catalyzes hydrogen peroxide-dependent oxidations and the formation of hypochlorous acid that is central to the microbicidal activity of netrophils.

Specificity

The immunogen sequence is present in the pro-fom and the mature forms of myeloperoxidase (without a.a. 1-48, a.a. 1-154, or a.a. 1-164), as well as the cleaved heavy chain (a.a. 279-745), but not light chain (a.a. 165-278).

Immunogen

KLH-conjugated linear peptide corresponding to a sequence within human myeloperoxidase heavy chain region.

Application

Anti-Myeloperoxidase Antibody, Cat. No. 07-496-I, is a highly specific rabbit polyclonal antibody that targets Myeloperoxidase and has been tested in Flow Cytometry and Western Blotting.
Flow Cytometry Analysis: 0.1 µg from a representative lot detected myeloperoxidase immunoreactivity in 4% paraformaldehyde-fixed, 0.2% Triton X-100-permeabilized HL-60 cells.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected myeloperoxidase heavy chain in 10 µg of Jurkat cell lysate.

Target description

~92/60 kDa (glycosylated pro-form/heavy chain) observed. 83.87/73.85/87.25 kDa (isoform H17/H14/H7 pro-form), 78.94/82.32 kDa (isoform H17/H7 with a.a. 1-48 propeptide removed), 66.45/66.45/69.83 kDa (isoform H17/H14/H7 with a.a. 1-154 removed), 65.48/65.48/68.86 kDa (isoform H17/H7 isoform H17/H14/H7 with a.a. 1-164 removed), 52.51 kDa (heavy chain; a.a. 279-745) calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 07-496

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leticija Zlatar et al.
Cell death & disease, 15(7), 548-548 (2024-08-01)
Tuberculosis (TB) remains one of the top 10 causes of death worldwide and still poses a serious challenge to public health. Recent attention to neutrophils has uncovered unexplored areas demanding further investigation. Therefore, the aim of this study was to
Tatiana Reshetnyak et al.
International journal of molecular sciences, 24(11) (2023-06-10)
Neutrophil Extracellular Traps (NETs) have been implicated in systemic lupus erythematosus (SLE) and antiphospholipid syndrome (APS) pathogenesis. The myeloperoxidase-deoxyribonucleic acid (MPO-DNA) complex and nucleosomes are serum markers of NETosis. The aim of this study was to assess these NETosis parameters
Hubert Hayden et al.
PloS one, 16(4), e0250265-e0250265 (2021-04-23)
Over the past years, neutrophil extracellular traps (NETs) were shown to contribute to states of acute and chronic inflammatory disease. They are composed of expelled chromatin and decorated by neutrophil-derived proteins. Therefore, the analysis of DNA complexes with myeloperoxidase (MPO)
Tina Uroda et al.
Nature protocols, 15(6), 2107-2139 (2020-05-27)
Long noncoding RNAs (lncRNAs) are recently discovered transcripts that regulate vital cellular processes, such as cellular differentiation and DNA replication, and are crucially connected to diseases. Although the 3D structures of lncRNAs are key determinants of their function, the unprecedented
Cristina de Diego et al.
Respiratory research, 25(1), 48-48 (2024-01-20)
Neutrophil extracellular traps (NETs) have repeatedly been related to COVID-19 severity and mortality. However, there is no consensus on their quantification, and there are scarce data on their evolution during the disease. We studied circulating NET markers in patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service