Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.46469

Millipore

Fluid D acc. USP

ready-to-use, bottle volume 500 mL , filling volume

Synonym(s):

Washing Fluid, Rinse Fluid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

USP 71

Quality Level

description

500 ml rinsing fluid in 500ml bottle with screw cap and septum; suitable for sterility testing

sterility

sterile; autoclaved

shelf life

12 mo.

feature

closure type screw cap with septum
ready-to-use

packaging

pkg of 500 mL
bottle of

technique(s)

sterile filtration: suitable

bottle capacity

500 mL

bottle volume

500 mL , filling volume

pH

7.1 ( in H2O)
7.1

application(s)

cosmetics

storage temp.

2-25°C

General description

Fluid D is a washing solution for sterility testing of pharmaceutical samples containing lecithin or oil using the membrane filtration method. Rinse the membrane with Fluid D after filtration and provide a suitable medium to observe microbial growth.

Application

Fluid D is suitable for rinsing the solutions containing oil or lecithin during sterility testing by membrane filtration.
The oil should penetrate the membrane by its own weight followed by filtering applying the pressure or suction gradually. The membrane may be washed 3 times with 100 ml Fluid D (Fluid A with 0.1% Tween® 80) afterwards. At least one or one half of the filter is incubated in Tryptic Soy Broth (e.g. article number 146458) and a second or the other half of the filter in Fluid Thioglycollate Medium (e.g. article number 146385) for not less than 14 days.

Features and Benefits

Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopeia requirements. Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests. Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, thus improving accuracy and reducing the risk of incorrect interpretation and false results.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service