Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.46452

Millipore

HAYFLICK Liquid Medium

suitable for bioburden testing, For mycoplasma detection

Synonym(s):

HAYFLICK Broth, Mycoplasma Broth, Mycoplasma Liquid Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
NACRES:
NA.75

Agency

EP 2.6.7
USP

Quality Level

sterility

sterile; aseptically filled

form

liquid

feature

ready-to-use

packaging

pkg of 100 mL (in 125 mL Plastic bottle with Screw cap)

pH

7.8 ( in H2O)

application(s)

bioburden testing
industrial qc
pharmaceutical

storage temp.

2-8°C

suitability

mycoplasma

General description

HAYFLICK Broth supplemented with horse serum provides useable cholesterol and fatty acids. Deoxyribonucleic acid provides purine and pyrimidine bases, which cannot be synthesized by mycoplasmas. A color change of the medium to yellow or purple indicates the growth of mycoplasmas due to the presence of phenol red. Penicillin inhibits the growth of accompanying bacterial flora.

Application

HAYFLICK Broth is used for the general detection of mycoplasmas in pharmaceutical products, especially in bulk vaccines, cell banks, and virus cultures.

Features and Benefits

  • Ready-to-use convenience as no further preparation steps are required.
  • Pharmaceutical products such as Biopharmaceuticals, Vaccines, Cell cultures, and Virus cultures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service