Skip to Content
MilliporeSigma

Skip To

1.46007

FRIIS Agar

according to EP 2.6.7, USP, plate diam. 60 mm, ready-to-use, Settle plate, sterile; aseptically filled, box of 20 plates, for mycoplasma

Synonym(s):

Mycoplasma Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.75
UNSPSC Code:
41106200
Plate diam.:
60 mm
Feature:
ready-to-use Settle plate, ready-to-use
Sterility:
sterile; aseptically filled
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

USP, according to EP 2.6.7

description

For non-avian mycoplasma detection

sterility

sterile; aseptically filled

form

plate

feature

ready-to-use Settle plate, ready-to-use

packaging

box of 20 plates

plate diam.

60 mm

pH

7.4 ( in H2O)

application(s)

industrial qc
microbiology
pharmaceutical

storage temp.

2-8°C

suitability

mycoplasma

General description

FRIIS Agar is supplemented with horse serum and swine serum to provide useable cholesterol and fatty acids. A color change of the medium to yellow or purple indicates the growth of mycoplasmas due to the presence of Phenol red. Bacitracin inhibits the synthesis of the cell wall of accompanying Gram-positive bacteria.

Application

FRIIS Agar is recommended for the detection of non-avian mycoplasmas in pharmaceutical products, especially in bulk vaccines, cell banks, and virus cultures.

Features and Benefits

  • Ready-to-use convenience as no further preparation steps are required.
  • Pharmaceutical products such as Biopharmaceuticals, Vaccines, Cell cultures, and Virus cultures.


Storage Class

11 - Combustible Solids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library