Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.46403

Millipore

Buffered Peptone Water

ReadyTube®, ISO 6887, ISO 6579, bottle capacity 1 L, bottle filling volume 1 L, closure type, Blue screw cap with 3 loci, box of 6 bottles

Synonym(s):

BPW, Peptone water (Buffered), ReadyTube® Buffered Peptone Water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

ISO 6579
ISO 6887

Quality Level

form

liquid

feature

closure type Blue screw cap with 3 loci
ready-to-use

packaging

box of 6 bottles

manufacturer/tradename

ReadyTube®

bottle capacity

1 L

bottle filling volume

1 L

pH

7.0 ( in H2O)

application(s)

food and beverages
industrial qc
pathogen testing
sample preparation

storage temp.

2-25°C

suitability

Salmonella spp.

General description

ReadyTube® is ready to use media to provide the highest level of quality and testing confidence for improving accuracy and significantly reducing the risk of false results. ReadyTube® buffered peptone water is a pre-enrichment medium designed to help the recovery of sub-lethally damaged Salmonellae before transfer to a selective medium. This pre-enrichment medium is free from inhibitors and is well buffered and provides conditions for resuscitation of the cells that have been injured by processes of food preservation. The media contains Peptone including an enzymatic digest of casein as a source of carbon, nitrogen, vitamins, and minerals. Sodium chloride maintains the osmotic balance and phosphates buffer the medium. The broth is rich in nutrients and produces high resuscitation rates for sub-lethally injured bacteria and supports intense growth.

Application

ReadyTube® buffered peptone water is used in preliminary non-selective enrichment of bacteria, particularly pathogenic Enterobacteriaceae such as Salmonella and Cronobacter, from food, animal feed, water and as diluent for sample preparation.

Legal Information

READYTUBE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service