Skip to Content
MilliporeSigma

1.37064

SAFC

Isopropyl-beta-D-thiogalactopyranoside

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51171641

Assay

≥99.0% (HPLC)

Quality Level

form

solid

mp

109-111 °C

transition temp

flash point >110 °C

solubility

10 g/L

storage temp.

2-8°C

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Application

Isopropyl-beta-D-thiogalactopyranoside. CAS 367-93-1, molar mass 238.3 g/mol.

Analysis Note

Assay (1H-NMR-content):≥ 95.0 %
Purity (HPLC):≥ 99.0 %
Identity:passes test
Color of solution (5 %, water):passes test
Turbidity of solution (0,5 %, water):passes test
Specific rotation (α 20/D; 1 mol/L; water):-32.5 - -30.5 °
Dioxane (HS-GC):≤ 380 ppm
Other residual solvents (ICH Q3C):excluded by the manufacturing process
Water (according to Karl Fischer):≤ 1.0 %

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service