Skip to Content
MilliporeSigma

420322

IPTG, Dioxane-Free, High Purity

CAS 367-93-1 is used in the stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function.

Synonym(s):

IPTG, Dioxane-Free, High Purity, Isopropyl-β-D-thiogalactopyranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H18O5S
CAS Number:
Molecular Weight:
238.30
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

impurities

≤0.1% Dioxane (GC)

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S([C@@H]1O[C@@H]([C@@H]([C@@H]([C@H]1O)O)O)CO)C(C)C

InChI

1S/C9H18O5S/c1-4(2)15-9-8(13)7(12)6(11)5(3-10)14-9/h4-13H,3H2,1-2H3/t5-,6+,7+,8-,9+/m1/s1

InChI key

BPHPUYQFMNQIOC-NXRLNHOXSA-N

General description

Inducer of β-D-galactosidase. Used in the stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function. IPTG induces the expression of cloned genes linked to the lac promoter.
See also IPTG, Animal-Free, High Purity, Cat. No. 420291.
Used in the stimulation of β-galactosidase in cellular systems in which dioxane would disrupt normal cell function. This preparation contains ≤0.1% dioxane by GC.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library