Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.24541

Millipore

Nicotinamide adenine dinucleotide phosphate disodium salt

pH 3.7-4.2 (10 mg/mL in water), ≥95.0% dry basis (enzymatic)

Synonym(s):

Nicotinamide adenine dinucleotide phosphate disodium salt, NADP disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7Na2O17P3
CAS Number:
Molecular Weight:
788.38
MDL number:
UNSPSC Code:
12352107
EC Index Number:
246-129-8
NACRES:
NA.21

Quality Level

Assay

≥95.0% dry basis (enzymatic)

form

solid

pH

3.7-4.2 (10 mg/mL in water)

storage temp.

2-8°C

SMILES string

[Na].[Na].[P](=O)([O-])(O[P](=O)(OC[C@H]3O[C@H]([C@@H]([C@@H]3O)O[P](=O)(O)O)[n]4c5ncnc(c5nc4)N)O)OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)[n+]2cc(ccc2)C(=O)N

InChI

1S/C21H28N7O17P3.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39)

InChI key

WINXVPJGUQYDJA-WUEGHLCSSA-N

General description

for biochemistry NADP-Na₂

Application


  • Biochemical and molecular modulation of CCl4-induced peripheral and central damage by Tilia americana var. mexicana extracts.: Investigates the protective biochemical pathways enhanced by NADP, addressing oxidative stress and tissue damage in clinical settings, suggesting therapeutic potentials in pharmacology (Coballase-Urrutia et al., 2017).

  • Selective Irreversible Inhibition of Neuronal and Inducible Nitric-oxide Synthase in the Combined Presence of Hydrogen Sulfide and Nitric Oxide.: Discusses the interaction of NADP in complex biochemical pathways influencing enzymatic activity, important for understanding cellular signaling and potential drug targets (Heine et al., 2015).

  • Treatment with n-3 polyunsaturated fatty acids reverses endothelial dysfunction and oxidative stress in experimental menopause.: This research outlines the critical roles of NADP in managing oxidative stress and cellular health, showcasing its implications in dietary supplements and therapeutic interventions (Gortan Cappellari et al., 2013).


Analysis Note

Assay (enzymatically; calculated on solvent-free substance): ≥ 95 %
Identity (IR-spectrum): passes test
pH (10 mg/ml; water): 3.7 - 4.2
UV-Absorption (360 nm; 10 mg/ml; water): ≤ 0.20
Spec. Absorptivity A 1%/1cm (260 nm; 0.01 g/l; buffer pH 7.0; calculated on solvent-free substance): 220 - 240
Methanol (GC): ≤ 3.0 %
Water (according to Karl Fischer): ≤ 6.0 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service