Skip to Content
MilliporeSigma

481972

NADP, Disodium Salt

A coenzyme that is widely distributed in living matter. Participates in oxidation-reduction reactions.

Synonym(s):

NADP, Disodium Salt, β-Nicotinamide-adenine Dinucleotide Phosphate, TPN, 2Na

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
41106305
NACRES:
NA.51
MDL number:
Assay:
≥95% (enzymatic)
Form:
solid
Solubility:
0.001% aqueous buffer, pH 7.0: soluble
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

Merck USA index - 14, 6348

assay

≥95% (enzymatic)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

0.001% aqueous buffer, pH 7.0: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Na].[Na].[P](=O)([O-])(O[P](=O)(OC[C@H]3O[C@H]([C@@H]([C@@H]3O)O[P](=O)(O)O)[n]4c5ncnc(c5nc4)N)O)OC[C@H]1O[C@H]([C@@H]([C@@H]1O)O)[n+]2cc(ccc2)C(=O)N

InChI key

WINXVPJGUQYDJA-WUEGHLCSSA-N

General description

A coenzyme that is widely distributed in living matter, participating in aerobic and anaerobic oxidations.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library