Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12-439

Sigma-Aldrich

Retinoblastoma Protein (aa 773-928)

Recombinant fusion protein containing the C-terminal fragment (residues 773-928) of human retinoblastoma protein (Rb) with an N-terminal His6-tag, expressed in E. coli.

Synonym(s):

Rb Protein, Retinoblastoma Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

recombinant

expressed in E. coli

Quality Level

form

liquid

manufacturer/tradename

Upstate®

concentration

2.0 mg/mL

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... RB1(5925)

Quality

Routinely evaluated as a sustrate for many Serine/Threonine kinases.

Physical form

50 mM Tris-HCl, pH 7.5, 150 mM NaCl, and 500 mM imidazole

Storage and Stability

Stable for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alberto Viera et al.
Journal of cell science, 128(1), 88-99 (2014-11-09)
In most organisms, telomeres attach to the nuclear envelope at the onset of meiosis to promote the crucial processes of pairing, recombination and synapsis during prophase I. This attachment of meiotic telomeres is mediated by the specific distribution of several
Huikun Zhang et al.
PLoS computational biology, 15(8), e1006813-e1006813 (2019-08-06)
Prediction of compounds that are active against a desired biological target is a common step in drug discovery efforts. Virtual screening methods seek some active-enriched fraction of a library for experimental testing. Where data are too scarce to train supervised
Michael J O'Connor et al.
Oncogene, 40(30), 4872-4883 (2021-06-24)
Cyclin D1 is an essential regulator of the G1-S cell-cycle transition and is overexpressed in many cancers. Expression of cyclin D1 is under tight cellular regulation that is controlled by many signaling pathways. Here we report that PARP14, a member
Severine Isabelle Gharbi et al.
NPJ breast cancer, 8(1), 126-126 (2022-11-30)
Despite the biological and therapeutic relevance of CDK4/6 for the treatment of HR+, HER2- advanced breast cancer, the detailed mode of action of CDK4/6 inhibitors is not completely understood. Of particular interest, phosphorylation of CDK4 at T172 (pT172) is critical
Petra Mikolcevic et al.
Nature communications, 7, 11084-11084 (2016-03-31)
Cyclin-dependent kinases (CDKs) play key roles in cell cycle regulation. Genetic analysis in mice has revealed an essential role for Cdk2 in meiosis, which renders Cdk2 knockout (KO) mice sterile. Here we show that mice deficient in RingoA, an atypical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service